Learn More
Alkali Scientific CellPro™ Low Evap 96-Well Cell Culture Plates, Flat Bottom, Sterile, Tissue Treated, 100/Cs
SDP
Catalog No. 503027552
Encompass_Preferred
CellPro™ now offers a family of polystyrene TC-treated or non-treated multi-well plates. These plates are optimized for general cell culture, robotics, plate readers, and liquid handling systems. Plates are available with various well sizes. CellPro™ cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assures uniform cell growth. Tissue culture-treated and non-treated are both available. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. Features: Available in individual packaging High clarity, 100% virgin polystyrene Sterilized by E-beam, SAL=10-6.
Catalog No. 50-302-7552 Supplier Alkali Scientific Supplier No. TPN1096TLEVP
CellPro™ now offers a family of polystyrene TC-treated or non-treated multi-well plates. These plates are optimized for general cell culture, robotics, plate readers, and liquid handling systems. Plates are available with various well sizes. CellPro™ cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assures uniform cell growth. Tissue culture-treated and non-treated are both available. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. Features: Available in individual packaging High clarity, 100% virgin polystyrene Sterilized by E-beam, SAL=10-6.