Learn More
Alkali Scientific CellPro™ Magnesium Acetate 1M 100mL
SDP
Catalog No. 502389394
Encompass_Preferred
Alkali Scientific™ CellPro™ Magnesium Acetate 1M, available in a 100mL volume, offers a meticulously prepared solution that meets the rigorous standards required for molecular biology and biochemical research. With its sterile and filtered formulation, this solution ensures optimal purity and reliability, essential for precise laboratory experiments. Magnesium acetate is a vital reagent utilized in various molecular biology applications, including nucleic acid purification, protein structure studies, and enzyme assays. The stringent quality control measures and precisely calibrated formulation of CellPro™ Magnesium Acetate guarantee consistent and reproducible results, empowering researchers to advance their scientific investigations with confidence. Trust in the purity and reliability of CellPro™ Magnesium Acetate to meet the demanding requirements of your laboratory's molecular biology and biochemical studies.
Catalog No. 50-238-9394 Supplier Alkali Scientific Supplier No. DB0239
Alkali Scientific™ CellPro™ Magnesium Acetate 1M, available in a 100mL volume, offers a meticulously prepared solution that meets the rigorous standards required for molecular biology and biochemical research. With its sterile and filtered formulation, this solution ensures optimal purity and reliability, essential for precise laboratory experiments. Magnesium acetate is a vital reagent utilized in various molecular biology applications, including nucleic acid purification, protein structure studies, and enzyme assays. The stringent quality control measures and precisely calibrated formulation of CellPro™ Magnesium Acetate guarantee consistent and reproducible results, empowering researchers to advance their scientific investigations with confidence. Trust in the purity and reliability of CellPro™ Magnesium Acetate to meet the demanding requirements of your laboratory's molecular biology and biochemical studies.