Learn More
Alkali Scientific CellPro™ Medium 199 with Earle's Balanced Salt Solution (EBSS), L-glutamine, 500ml, 10/Case
SDP
Catalog No. 503028817
Encompass_Preferred
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 50-302-8817 Supplier Alkali Scientific Supplier No. M199CS
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.