Learn More
Alkali Scientific CellPro™ MEM Amino Acid solution, 100ml, 10/Case
SDP
Catalog No. 503027407
Encompass_Preferred
  Growth supplement for cell culture medium Supplement for serum- and protein-free media. Support the growth of cells that have high energy demands and synthesize large amounts of proteins and nucleic acids
Catalog No. 50-302-7407 Supplier Alkali Scientific Supplier No. MEM504CS
  Growth supplement for cell culture medium Supplement for serum- and protein-free media. Support the growth of cells that have high energy demands and synthesize large amounts of proteins and nucleic acids