Learn More
Alkali Scientific CellPro MEM with EBSS L-glutam
Supplier: Alkali Scientific MEM500
MEM liquid media supports cell growth in research upstream process development and cell culture manufacturing processes Minimum essential medium efficacy and homogeneity is tested MEM composition includes vitamins minerals and amino acids vital to supporting mammalian cell culture and promoting cell growth There are several MEM media options to support cell culture process development Formulations of cell culture MEM media are available with or without L-glutamine The MEM alpha modification includes the additional components such as non-essential amino acids ascorbic acid biotin vitamin B12 lipoic acid sodium pyruvate ribonucleosides and deoxyribonucleosides Improved MEM or Richters modification is also available with increased concentrations of certain amino acids vitamins and metabolites In many MEM media formulations phenol red is added as a pH indicator All available MEM media contain a sodium bicarbonate buffer with some also containing EBSS to maintain optimal culture pH