Learn More
Alkali Scientific CellPro™ Microtube w/ cap, 0.5mL, opaque black, 500/pack
SDP
Catalog No. 503026400
Encompass_Preferred
Our CellPro™ Microtube are made of transparent polymer material polypropylene (PP), the tube cover lock design can provide better sealing and sample protection, which can effectively avoid accidental opening of the cover and prevent evaporation of samples during long-term storage, ensuring operational safety.  500/pack
Catalog No. 50-302-6400 Supplier Alkali Scientific Supplier No. C3006OB
Our CellPro™ Microtube are made of transparent polymer material polypropylene (PP), the tube cover lock design can provide better sealing and sample protection, which can effectively avoid accidental opening of the cover and prevent evaporation of samples during long-term storage, ensuring operational safety.  500/pack