Learn More
Alkali Scientific CellPro™ MOPS, 10X, Brown Plastic Bottle, 1L, 12/cs
SDP
Catalog No. 502389349
Encompass_Preferred
Alkali Scientific™ CellPro™ MOPS Buffer 10X is a versatile reagent commonly used in molecular biology and biochemistry applications. With its optimized formulation, this buffer provides stable pH conditions, crucial for nucleic acid electrophoresis, RNA isolation, and protein gel electrophoresis. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ MOPS Buffer 10X a trusted choice for researchers seeking reliable results in their experiments.
Catalog No. 50-238-9349 Supplier Alkali Scientific Supplier No. DB0356CS
Add to Cart
Add to Cart
Alkali Scientific™ CellPro™ MOPS Buffer 10X is a versatile reagent commonly used in molecular biology and biochemistry applications. With its optimized formulation, this buffer provides stable pH conditions, crucial for nucleic acid electrophoresis, RNA isolation, and protein gel electrophoresis. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ MOPS Buffer 10X a trusted choice for researchers seeking reliable results in their experiments.