Learn More
Alkali Scientific CellPro™ Multi Insert Dish, Sterile, External Grip, 15mL, 60mm x 20mm, 18/Cs, Low Binding Treatment, Non Groove Bottom, Untreated
SDP
Catalog No. 5000662166
Encompass_Preferred
The CellPro™ Multi Insert Dish is a sterile 15mL dish designed for cell culture, featuring a non-groove bottom and low binding treatment. The 60mm x 20mm dish is perfect for studying cell behaviors and interactions in laboratory experiments. With an external grip for easy handling and 18 dishes per case, this product ensures convenience and high-quality results in research applications. It is ideal for researchers requiring multi-insert solutions.
Catalog No. 50-006-62166 Supplier Alkali Scientific Supplier No. MID0025
The CellPro™ Multi Insert Dish is a sterile 15mL dish designed for cell culture, featuring a non-groove bottom and low binding treatment. The 60mm x 20mm dish is perfect for studying cell behaviors and interactions in laboratory experiments. With an external grip for easy handling and 18 dishes per case, this product ensures convenience and high-quality results in research applications. It is ideal for researchers requiring multi-insert solutions.