Learn More
Alkali Scientific CellPro™ Multi Purpose Sealing Film, 1.2" Wide, 125'/ Roll, RNase, DNase, Pyrogen Free
SDP
Catalog No. 5000662719
Encompass_Preferred
HyperStretch™ Sealing Film is a highly elastic laboratory sealing film developed to provide dependable sealing for a wide range of laboratory containers. Its exceptional stretchability allows the film to conform tightly around vessels of varying shapes and s, including flasks, tubes, beakers, and culture containers. This secure seal helps protect samples from contamination, evaporation, and accidental spills during handling, storage, or transport. Manufactured to meet laboratory quality standards, the film is RNase, DNase, and pyrogen free, supporting sensitive research and molecular biology applications. Easy to apply and remove without leaving adhesive residue, this versatile sealing film is well suited for research laboratories, clinical environments, and routine laboratory workflows.
Catalog No. 50-006-62719 Supplier Alkali Scientific Supplier No. SF0389
HyperStretch™ Sealing Film is a highly elastic laboratory sealing film developed to provide dependable sealing for a wide range of laboratory containers. Its exceptional stretchability allows the film to conform tightly around vessels of varying shapes and s, including flasks, tubes, beakers, and culture containers. This secure seal helps protect samples from contamination, evaporation, and accidental spills during handling, storage, or transport. Manufactured to meet laboratory quality standards, the film is RNase, DNase, and pyrogen free, supporting sensitive research and molecular biology applications. Easy to apply and remove without leaving adhesive residue, this versatile sealing film is well suited for research laboratories, clinical environments, and routine laboratory workflows.