Learn More
Alkali Scientific CellPro™ Multi Purpose Sealing Film, 2" Wide, 125'/ Roll, RNase, DNase, Pyrogen Free
SDP
Catalog No. 5000663119
Encompass_Preferred
HyperStretch™ Sealing Film is a highly stretchable laboratory film designed to form a reliable, leak resistant seal on a wide variety of labware. The elastic material easily conforms to containers of different shapes and s, including tubes, flasks, beakers, and culture vessels, helping protect samples from contamination, evaporation, and accidental spills. Its flexible design supports dependable sealing during sample storage, transport, and everyday laboratory handling. Manufactured to meet laboratory standards, the film is RNase, DNase, and pyrogen free, making it suitable for sensitive molecular and analytical applications. Easy to apply and remove without leaving residue, this multi purpose sealing film is ideal for research labs, teaching facilities, and clinical laboratories.
Catalog No. 50-006-63119 Supplier Alkali Scientific Supplier No. SF0385
HyperStretch™ Sealing Film is a highly stretchable laboratory film designed to form a reliable, leak resistant seal on a wide variety of labware. The elastic material easily conforms to containers of different shapes and s, including tubes, flasks, beakers, and culture vessels, helping protect samples from contamination, evaporation, and accidental spills. Its flexible design supports dependable sealing during sample storage, transport, and everyday laboratory handling. Manufactured to meet laboratory standards, the film is RNase, DNase, and pyrogen free, making it suitable for sensitive molecular and analytical applications. Easy to apply and remove without leaving residue, this multi purpose sealing film is ideal for research labs, teaching facilities, and clinical laboratories.