Learn More
Alkali Scientific CellPro™ Multi Purpose Sealing Film, 4 x 4" Pre Cut Sheets, 100/Pk, Sterile
SDP
Catalog No. 5000662910
Encompass_Preferred
HyperStretch™ Sealing Film is a highly elastic laboratory sealing film designed to create a secure, leak resistant seal on a wide variety of labware. Its exceptional stretchability allows it to conform tightly to irregular shapes and containers of different s, helping protect valuable samples from contamination, evaporation, and accidental spills. The flexible film stretches easily over flasks, beakers, tubes, and other laboratory vessels while maintaining a dependable seal during routine handling, storage, or transport. Easy to apply and remove, it leaves no sticky residue and supports efficient laboratory workflows. This multi purpose sealing film is ideal for research laboratories, clinical environments, teaching labs, and quality control settings where reliable sample protection and containment are essential.
Catalog No. 50-006-62910 Supplier Alkali Scientific Supplier No. SF0400
HyperStretch™ Sealing Film is a highly elastic laboratory sealing film designed to create a secure, leak resistant seal on a wide variety of labware. Its exceptional stretchability allows it to conform tightly to irregular shapes and containers of different s, helping protect valuable samples from contamination, evaporation, and accidental spills. The flexible film stretches easily over flasks, beakers, tubes, and other laboratory vessels while maintaining a dependable seal during routine handling, storage, or transport. Easy to apply and remove, it leaves no sticky residue and supports efficient laboratory workflows. This multi purpose sealing film is ideal for research laboratories, clinical environments, teaching labs, and quality control settings where reliable sample protection and containment are essential.