Learn More
Alkali Scientific CellPro™ Multichannel Solution Reservoir, 12-Channel, 48mL, Holds up to 12 Reagents, (12 x 4.0mL), Sterile, PS, Individually Wrapped, 100/Cs
SDP
Catalog No. 503028216
Encompass_Preferred
These disposable polystyrene reservoirs are ideal for handling multiple solutions at once. Designed for use with multichannel pipettors, they are also useful on automated and semi-automated platforms and in other high-throughput applications. Available in an 8-channel or 12-channel configuration, these reservoirs are individually wrapped sterile in easy to open bags, and certified RNAse, DNase and pyrogen free. • 8 or 12 Channel Configurations• Compatible with Multichannel Pipettors• Individually Wrapped in Easy Tear Bags• Sterile; RNase, DNase, and Pyrogen Free
Catalog No. 50-302-8216 Supplier Alkali Scientific Supplier No. RS4812
These disposable polystyrene reservoirs are ideal for handling multiple solutions at once. Designed for use with multichannel pipettors, they are also useful on automated and semi-automated platforms and in other high-throughput applications. Available in an 8-channel or 12-channel configuration, these reservoirs are individually wrapped sterile in easy to open bags, and certified RNAse, DNase and pyrogen free. • 8 or 12 Channel Configurations• Compatible with Multichannel Pipettors• Individually Wrapped in Easy Tear Bags• Sterile; RNase, DNase, and Pyrogen Free