Learn More
Alkali Scientific CellPro™ Neutralizing Solution, 4L Cubitainer
SDP
Catalog No. 502389172
Encompass_Preferred
CellPro™ Neutralizing Solution is a specialized formula designed to effectively neutralize trypsin and other enzymatic solutions used in cell culture. It protects delicate cells during dissociation and ensures optimal recovery by maintaining cell viability. Ideal for lab applications, it is sterile, ready-to-use, and supports consistent cell passaging and subculturing processes.
Catalog No. 50-238-9172 Supplier Alkali Scientific Supplier No. DS02154L
Add to Cart
Add to Cart
CellPro™ Neutralizing Solution is a specialized formula designed to effectively neutralize trypsin and other enzymatic solutions used in cell culture. It protects delicate cells during dissociation and ensures optimal recovery by maintaining cell viability. Ideal for lab applications, it is sterile, ready-to-use, and supports consistent cell passaging and subculturing processes.