Learn More
Alkali Scientific CellPro™ Newborn Calf (Bovine) Serum, US Source, 500mL
SDP
Catalog No. 503026891
Encompass_Preferred
Derived from healthy animals up to 20 days of age, that has no known reportable diseases. Newborn Calf Serum from contains higher protein and IgG levels when compared to Fetal Bovine Serum. NBCS may be used as an alternative to more costly FBS in certain cell culture applications. Produced from raw materials collected from approved facilities within the contiguous United States that has undergone inspection by the United States Department of Agriculture. Manufactured utilizing current Good Manufacturing Practices (cGMP). 0.2 micron filtered. Average endotoxin level*: < 2 EU/ml (specification = ≤ 35 EU/ml) Average hemoglobin level*: < 20 mg/dl (specification = < 30 mg/dL) 9CFR tested 0.2 µm filtered US origin Storage conditions: ≤ -10° C
Catalog No. 50-302-6891 Supplier Alkali Scientific Supplier No. FB56
Derived from healthy animals up to 20 days of age, that has no known reportable diseases. Newborn Calf Serum from contains higher protein and IgG levels when compared to Fetal Bovine Serum. NBCS may be used as an alternative to more costly FBS in certain cell culture applications. Produced from raw materials collected from approved facilities within the contiguous United States that has undergone inspection by the United States Department of Agriculture. Manufactured utilizing current Good Manufacturing Practices (cGMP). 0.2 micron filtered. Average endotoxin level*: < 2 EU/ml (specification = ≤ 35 EU/ml) Average hemoglobin level*: < 20 mg/dl (specification = < 30 mg/dL) 9CFR tested 0.2 µm filtered US origin Storage conditions: ≤ -10° C