Learn More
Alkali Scientific CellPro Nucleic Maxiprep Purif
SDP
Catalog No. 503029010
Encompass_Preferred
The Alkali Scientific Nucleic Acid Maxiprep Purification Column is designed for the efficient extraction and purification of nucleic acids making it an essential tool for molecular biology genetic engineering and diagnostic research This high-performance column enables large-scale purification of DNA and RNA delivering high yields with exceptional purity and consistency Built with a medical-grade six-layer glass fiber membrane this purification column ensures strong binding efficiency structural durability and reliable performance Its advanced filtration system optimizes nucleic acid recovery making it ideal for applications such as PCR qPCR sequencing cloning hybridization and gene expression analysis Six-Layer Filtration High-quality glass fiber membrane enhances nucleic acid binding and purification High Yield and Purity Efficiently isolates large quantities of DNA or RNA with minimal contaminants Large Processing Volume Features a 20 mL purification column for high-throughpu
Catalog No. 50-302-9010 Supplier Alkali Scientific Supplier No. NAPC001
The Alkali Scientific Nucleic Acid Maxiprep Purification Column is designed for the efficient extraction and purification of nucleic acids making it an essential tool for molecular biology genetic engineering and diagnostic research This high-performance column enables large-scale purification of DNA and RNA delivering high yields with exceptional purity and consistency Built with a medical-grade six-layer glass fiber membrane this purification column ensures strong binding efficiency structural durability and reliable performance Its advanced filtration system optimizes nucleic acid recovery making it ideal for applications such as PCR qPCR sequencing cloning hybridization and gene expression analysis Six-Layer Filtration High-quality glass fiber membrane enhances nucleic acid binding and purification High Yield and Purity Efficiently isolates large quantities of DNA or RNA with minimal contaminants Large Processing Volume Features a 20 mL purification column for high-throughpu