Learn More
Alkali Scientific CellPro™ NZCYM Media + 0.7% Top Agar
SDP
Catalog No. 502389119
Encompass_Preferred
Alkali Scientific™ CellPro™ NZCYM Media is a nutrient-rich growth medium optimized for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZCYM Media ensures purity and consistency, empowering researchers with reliable results in their molecular biology workflows.
Catalog No. 50-238-9119 Supplier Alkali Scientific Supplier No. DC0006
Alkali Scientific™ CellPro™ NZCYM Media is a nutrient-rich growth medium optimized for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZCYM Media ensures purity and consistency, empowering researchers with reliable results in their molecular biology workflows.