Learn More
Alkali Scientific CellPro™ NZCYM Media 500mL PC
SDP
Catalog No. 502388935
Encompass_Preferred
Alkali Scientific™ CellPro™ NZCYM Media is a nutrient-rich growth medium optimized for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZCYM Media ensures purity and consistency, empowering researchers with reliable results in their molecular biology workflows.
Catalog No. 50-238-8935 Supplier Alkali Scientific Supplier No. DC0020
Add to Cart
Add to Cart
Alkali Scientific™ CellPro™ NZCYM Media is a nutrient-rich growth medium optimized for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZCYM Media ensures purity and consistency, empowering researchers with reliable results in their molecular biology workflows.