Learn More
Alkali Scientific CellPro™ NZY Agar 150mm
SDP
Catalog No. 502388685
Encompass_Preferred
CellPro™ NZY Agar is a nutrient-rich medium used for the cultivation and selection of bacterial strains in microbiology and molecular biology applications. NZY Agar is a modified variant of LB (Luria-Bertani) agar, supplemented with yeast extract, peptone, sodium chloride, and agar. This agar medium provides essential nutrients for bacterial growth and is particularly suitable for the propagation of recombinant E. coli strains harboring plasmids with antibiotic resistance markers. The 150mm plates come in a pack of 10, providing a convenient solution for bacterial culture experiments.
Catalog No. 50-238-8685 Supplier Alkali Scientific Supplier No. DC42237
CellPro™ NZY Agar is a nutrient-rich medium used for the cultivation and selection of bacterial strains in microbiology and molecular biology applications. NZY Agar is a modified variant of LB (Luria-Bertani) agar, supplemented with yeast extract, peptone, sodium chloride, and agar. This agar medium provides essential nutrients for bacterial growth and is particularly suitable for the propagation of recombinant E. coli strains harboring plasmids with antibiotic resistance markers. The 150mm plates come in a pack of 10, providing a convenient solution for bacterial culture experiments.