Learn More
Alkali Scientific CellPro™ NZY Broth 1L
SDP
Catalog No. 502389486
Encompass_Preferred
Alkali Scientific™ CellPro™ NZY Broth is a nutrient-rich growth medium designed for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZY Broth ensures purity and consistency, empowering researchers with reliable results in their molecular biology workflows.
Catalog No. 50-238-9486 Supplier Alkali Scientific Supplier No. DC0029
Alkali Scientific™ CellPro™ NZY Broth is a nutrient-rich growth medium designed for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZY Broth ensures purity and consistency, empowering researchers with reliable results in their molecular biology workflows.