Learn More
Alkali Scientific CellPro™ NZY Media + 1.5% Agar, 500mL
SDP
Catalog No. 502389463
Encompass_Preferred
Alkali Scientific™ CellPro™ NZY Media is a nutrient-rich growth medium optimized for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZY Media ensures purity and consistency, empowering researchers with reliable results in their molecular biology workflows.
Catalog No. 50-238-9463 Supplier Alkali Scientific Supplier No. DC0031
Alkali Scientific™ CellPro™ NZY Media is a nutrient-rich growth medium optimized for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZY Media ensures purity and consistency, empowering researchers with reliable results in their molecular biology workflows.