Learn More
Alkali Scientific CellPro Oak Ridge Style High-S
SDP
Supplier:  Alkali Scientific OR10PPCOCS
Encompass_Preferred
CellProOak Ridge High-Speed PPCO Round Bottom Centrifuge Tubes with Attached Screw Caps are designed to be chemically compatible with a broader range of laboratory reagents than traditional polycarbonate centrifuge tubes and can withstand repeated autoclaving for prolonged use Enhanced Chemical Resistance Offers superior resistance compared to polycarbonate Durable Under Repeated Autoclaving Engineered to endure multiple autoclave cycles ensuring a longer lifespan than polycarbonate tubes High-Speed Performance When used with a CellPro sealing cap (sold separately) 30 mL tubes can be centrifuged at speeds up to 50000 x g while 50 mL tubes can reach up to 100605 x g User-Friendly Closures Equipped with polypropylene linerless closures that are easy to clean and provide leakproof protection under normal conditions up to 10000 x g Optimal Filling Requirement Tubes should be filled to at least 80% capacity for proper performance Autoclavable and Leakproof Remains autoclavable and lea
Catalog No. 50-302-7634