Learn More
Alkali Scientific CellPro™ PBS 10X, pH 7.0, 500mL, 500mL, 10/case
SDP
Catalog No. 503029092
Encompass_Preferred
CellPro™ PBS 10X, pH 7.0, 500mL Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.  
Catalog No. 50-302-9092 Supplier Alkali Scientific Supplier No. PB5970CS
Add to Cart
Add to Cart
CellPro™ PBS 10X, pH 7.0, 500mL Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.  