Learn More
Alkali Scientific CellPro™ PBS 10X, pH 7.2, without calcium, magnesium, 500mL, 500ml, 10/case
SDP
Catalog No. 503029091
Encompass_Preferred
CellPro™ PBS, 10X, pH7.2, 500mL Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.  
Catalog No. 50-302-9091 Supplier Alkali Scientific Supplier No. PB510CS
Add to Cart
Add to Cart
CellPro™ PBS, 10X, pH7.2, 500mL Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.  