Learn More
Alkali Scientific CellPro™ PBS Buffer, 1X, p.H 7.2, 1L, 1 Liter
SDP
Catalog No. 503027631
Encompass_Preferred
CellPro™ PBS Buffer, 1X pH 7.2, 1 Liter Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.
Catalog No. 50-302-7631 Supplier Alkali Scientific Supplier No. PB1000
CellPro™ PBS Buffer, 1X pH 7.2, 1 Liter Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.