Learn More
Alkali Scientific CellPro™ PBS Buffer, 1X, pH 7.2, 500mL
SDP
Catalog No. 502389339
Encompass_Preferred
Alkali Scientific™ CellPro™ PBS (Phosphate Buffered Saline) provides researchers with a reliable and consistent solution essential for various molecular and cellular biology applications. This meticulously prepared buffer is formulated to maintain a stable pH and osmolarity, ensuring optimal conditions for cell culture, protein assays, immunohistochemistry, and many other biological techniques. With its phosphate-based formulation, CellPro™ PBS offers excellent buffering capacity, making it suitable for maintaining the physiological pH of biological samples and preserving their integrity. Whether used for cell washing, sample dilution, or buffer exchange, CellPro™ PBS guarantees exceptional performance and reproducibility in every experiment. Trust in the quality and reliability of CellPro™ PBS to support your research needs and enable precise and consistent results.
Catalog No. 50-238-9339 Supplier Alkali Scientific Supplier No. PB501
Alkali Scientific™ CellPro™ PBS (Phosphate Buffered Saline) provides researchers with a reliable and consistent solution essential for various molecular and cellular biology applications. This meticulously prepared buffer is formulated to maintain a stable pH and osmolarity, ensuring optimal conditions for cell culture, protein assays, immunohistochemistry, and many other biological techniques. With its phosphate-based formulation, CellPro™ PBS offers excellent buffering capacity, making it suitable for maintaining the physiological pH of biological samples and preserving their integrity. Whether used for cell washing, sample dilution, or buffer exchange, CellPro™ PBS guarantees exceptional performance and reproducibility in every experiment. Trust in the quality and reliability of CellPro™ PBS to support your research needs and enable precise and consistent results.