Learn More
Alkali Scientific CellPro™ PBS Buffer, 1X, pH 7.4, without Calcium or Magnesium, Sterile Filtered, 1000mL, 10/Case
SDP
Catalog No. 503028332
Encompass_Preferred
CellPro™ PBS Buffer, 1X, pH 7.4, 1000mL Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.
Catalog No. 50-302-8332 Supplier Alkali Scientific Supplier No. PB1001CS
CellPro™ PBS Buffer, 1X, pH 7.4, 1000mL Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.