Learn More
Alkali Scientific CellPro™ Penicillin, Streptomycin, and Glutamine Solution, 100ml
SDP
Catalog No. 503029139
Encompass_Preferred
Effective against gram negative and gram positive microorganisms. Penicillin interferes with the final stage of synthesis of the bacterial cell wall. Streptomycin inhibits elongation at transpeptidation step during protein synthesis and binds to 30S subunit, causing misreading. Common working concentration for penicillin is 100 U/mL; for Streptomycin: 100 μg/mL. Penicillin/Streptomycin is a broad spectrum bacteriostatic with activity against gram negative and gram-positive organisms. ****This item is required to ship on dry ice. Overnight Shipping required, For Research Use Only (RUO)  
Catalog No. 50-302-9139 Supplier Alkali Scientific Supplier No. FB83
Effective against gram negative and gram positive microorganisms. Penicillin interferes with the final stage of synthesis of the bacterial cell wall. Streptomycin inhibits elongation at transpeptidation step during protein synthesis and binds to 30S subunit, causing misreading. Common working concentration for penicillin is 100 U/mL; for Streptomycin: 100 μg/mL. Penicillin/Streptomycin is a broad spectrum bacteriostatic with activity against gram negative and gram-positive organisms. ****This item is required to ship on dry ice. Overnight Shipping required, For Research Use Only (RUO)  