Learn More
Alkali Scientific CellPro™ Pestles for FlowTubes
SDP
Catalog No. 503028598
Encompass_Preferred
CellPro™ Pestles for FlowTubes for 35µm strainer caps, sterile, individually wrapped 100/Cs
Catalog No. 50-302-8598 Supplier Alkali Scientific Supplier No. CT6411
CellPro™ Pestles for FlowTubes for 35µm strainer caps, sterile, individually wrapped 100/Cs