Learn More
Alkali Scientific CellPro™ PET Media Bottles (Square) with HDPE Caps, 60mL, 48/Case
SDP
Catalog No. 503026381
Encompass_Preferred
CellPro square media bottles, composed of high-strength plastic material, are intended for storing and transporting general reagents and cell culture reagents.The square-shaped bottles are designed to maximize space saving, durability, strength, and clarity as close to glass as possible.The bottles are provided as 125mL, 250mL, 500mL, and 1000mL, and are shrink-wrapped in a double-walled corrugated paper boxes and thoroughly sterilized for best quality.  ▪ Molded-in graduation scales▪ Shrink-wrap tray modules▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Non-hemolytic▪ Human DNA – free
Catalog No. 50-302-6381 Supplier Alkali Scientific Supplier No. VB60
CellPro square media bottles, composed of high-strength plastic material, are intended for storing and transporting general reagents and cell culture reagents.The square-shaped bottles are designed to maximize space saving, durability, strength, and clarity as close to glass as possible.The bottles are provided as 125mL, 250mL, 500mL, and 1000mL, and are shrink-wrapped in a double-walled corrugated paper boxes and thoroughly sterilized for best quality.  ▪ Molded-in graduation scales▪ Shrink-wrap tray modules▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Non-hemolytic▪ Human DNA – free