Learn More
Alkali Scientific CellPro Plasmid Maxiprep Purif
SDP
Catalog No. 503028952
Encompass_Preferred
The Alkali Scientific Plasmid Maxiprep Purification Column is designed for the efficient isolation and purification of plasmid DNA making it an essential tool for molecular biology and genetic engineering research This high-performance column streamlines large-scale plasmid purification delivering over 1000 g of high-quality DNA with speed and reliability Constructed with a medical-grade six-layer glass fiber membrane this purification column ensures exceptional binding efficiency structural durability and consistent performance Its advanced filtration system enhances DNA yield and purity making it an ideal solution for researchers working with enzyme digestion transformation PCR amplification sequencing and library construction Six-Layer Filtration High-quality glass fiber membrane maximizes DNA binding and purification efficiency Large Processing Volume Features a 20 mL purification column for high-yield plasmid isolation Robust Design Built for impact resistance and reliable
Catalog No. 50-302-8952 Supplier Alkali Scientific Supplier No. PPC001
The Alkali Scientific Plasmid Maxiprep Purification Column is designed for the efficient isolation and purification of plasmid DNA making it an essential tool for molecular biology and genetic engineering research This high-performance column streamlines large-scale plasmid purification delivering over 1000 g of high-quality DNA with speed and reliability Constructed with a medical-grade six-layer glass fiber membrane this purification column ensures exceptional binding efficiency structural durability and consistent performance Its advanced filtration system enhances DNA yield and purity making it an ideal solution for researchers working with enzyme digestion transformation PCR amplification sequencing and library construction Six-Layer Filtration High-quality glass fiber membrane maximizes DNA binding and purification efficiency Large Processing Volume Features a 20 mL purification column for high-yield plasmid isolation Robust Design Built for impact resistance and reliable