Learn More
Alkali Scientific CellPro Polyester Biopsy Bags
SDP
Catalog No. 503026419
Encompass_Preferred
Optimize specimen processing and preservation with CellProPolyester Biopsy Bags designed for solvent and acid resistance with a durable synthetic mesh (0 2mm) These high-quality biopsy bags provide a secure and efficient solution for histological applications ensuring maximum tissue safety and seamless fluid exchange Available in three sizes30 x 45mm 45 x 75mm and 75 x 95mmto accommodate a range of sample types Solvent and acid-resistant polyester material for enhanced durability 0 2mm synthetic mesh allows optimal fluid exchange for thorough specimen processing Secure tissue containment prevents sample loss during fixation and processing Three size options to fit various specimen needs CellProbiopsy bags offer a practical and efficient method for handling biological specimens Simply pour the fixative and sample directly into the bag where the mesh design ensures effective filtration while securely containing the biopsy for accurate histological analysis Once sealed the biopsy bag
Catalog No. 50-302-6419 Supplier Alkali Scientific Supplier No. BBP003
Optimize specimen processing and preservation with CellProPolyester Biopsy Bags designed for solvent and acid resistance with a durable synthetic mesh (0 2mm) These high-quality biopsy bags provide a secure and efficient solution for histological applications ensuring maximum tissue safety and seamless fluid exchange Available in three sizes30 x 45mm 45 x 75mm and 75 x 95mmto accommodate a range of sample types Solvent and acid-resistant polyester material for enhanced durability 0 2mm synthetic mesh allows optimal fluid exchange for thorough specimen processing Secure tissue containment prevents sample loss during fixation and processing Three size options to fit various specimen needs CellProbiopsy bags offer a practical and efficient method for handling biological specimens Simply pour the fixative and sample directly into the bag where the mesh design ensures effective filtration while securely containing the biopsy for accurate histological analysis Once sealed the biopsy bag