Learn More
Alkali Scientific CellPro Polypropylene PP Centr
SDP
Catalog No. 503026401
Encompass_Preferred
Efficiently process large-volume cell harvesting pelleting and protein purification with CellProPolypropylene (PP) Centrifuge Bottlesa reliable solution engineered for demanding laboratory applications These bottles are crafted from high-quality opaque PP offering exceptional chemical resistance durability and compatibility with a wide range of laboratory reagents Their autoclavable nature ensures they can withstand rigorous sterilization processes making them ideal for high-performance workflows The opaque design of the bottles provides superior chemical resistance making them well-suited for applications involving aggressive solvents acids or bases While they do not allow for direct visual monitoring of samples like polycarbonate or PPCO their enhanced durability and resistance to autoclaving cycles make them a preferred choice for long-term use and high-stress applications To ensure optimal performance these centrifuge bottles must be filled to at least 80% of their total capac
Catalog No. 50-302-6401 Supplier Alkali Scientific Supplier No. CB1000PP
Efficiently process large-volume cell harvesting pelleting and protein purification with CellProPolypropylene (PP) Centrifuge Bottlesa reliable solution engineered for demanding laboratory applications These bottles are crafted from high-quality opaque PP offering exceptional chemical resistance durability and compatibility with a wide range of laboratory reagents Their autoclavable nature ensures they can withstand rigorous sterilization processes making them ideal for high-performance workflows The opaque design of the bottles provides superior chemical resistance making them well-suited for applications involving aggressive solvents acids or bases While they do not allow for direct visual monitoring of samples like polycarbonate or PPCO their enhanced durability and resistance to autoclaving cycles make them a preferred choice for long-term use and high-stress applications To ensure optimal performance these centrifuge bottles must be filled to at least 80% of their total capac