Learn More
Alkali Scientific CellPro™ Potassium Acetate 1M pH 7.5 100mL
SDP
Catalog No. 502389493
Encompass_Preferred
Alkali Scientific™ CellPro™ Potassium Acetate, Sterile, Filtered, offers exceptional purity and reliability for a variety of molecular biology and biochemical applications. This sterile and filtered potassium acetate solution is meticulously prepared to ensure minimal contamination, providing researchers with a high-quality reagent for their experimental needs. Potassium acetate is commonly used in molecular biology protocols for DNA and RNA precipitation, protein crystallization, and buffer preparation. With its stringent quality control measures and superior performance, CellPro™ Potassium Acetate is the ideal choice for laboratories requiring consistent and reliable results in their research endeavors. Trust in the purity and effectiveness of CellPro™ Potassium Acetate to support your scientific work with precision and confidence.
Catalog No. 50-238-9493 Supplier Alkali Scientific Supplier No. DB0235
Alkali Scientific™ CellPro™ Potassium Acetate, Sterile, Filtered, offers exceptional purity and reliability for a variety of molecular biology and biochemical applications. This sterile and filtered potassium acetate solution is meticulously prepared to ensure minimal contamination, providing researchers with a high-quality reagent for their experimental needs. Potassium acetate is commonly used in molecular biology protocols for DNA and RNA precipitation, protein crystallization, and buffer preparation. With its stringent quality control measures and superior performance, CellPro™ Potassium Acetate is the ideal choice for laboratories requiring consistent and reliable results in their research endeavors. Trust in the purity and effectiveness of CellPro™ Potassium Acetate to support your scientific work with precision and confidence.