Learn More
Alkali Scientific CellPro PPCO Centrifuge Bottle
SDP
Catalog No. 503026583
Encompass_Preferred
Efficiently process large-volume cell harvesting pelleting and protein purification with CellProPolypropylene Copolymer (PPCO) Centrifuge Bottlesan advanced solution engineered for high-performance laboratory applications These bottles are crafted from high-quality translucent PPCO offering enhanced chemical resistance durability and reduced risk of sample contamination Their autoclavable nature ensures they can withstand rigorous sterilization processes making them ideal for demanding workflows The translucent design allows for basic visual monitoring of sample contents which is particularly useful during centrifugation PPCO provides greater resistance to chemicals and temperature variations compared to polycarbonate making these bottles an excellent choice for applications involving aggressive solvents or extended storage conditions To ensure optimal performance these centrifuge bottles must be filled to at least 80% of their total capacity for proper balance and reliable centri
Catalog No. 50-302-6583 Supplier Alkali Scientific Supplier No. CB0500PPCO
Efficiently process large-volume cell harvesting pelleting and protein purification with CellProPolypropylene Copolymer (PPCO) Centrifuge Bottlesan advanced solution engineered for high-performance laboratory applications These bottles are crafted from high-quality translucent PPCO offering enhanced chemical resistance durability and reduced risk of sample contamination Their autoclavable nature ensures they can withstand rigorous sterilization processes making them ideal for demanding workflows The translucent design allows for basic visual monitoring of sample contents which is particularly useful during centrifugation PPCO provides greater resistance to chemicals and temperature variations compared to polycarbonate making these bottles an excellent choice for applications involving aggressive solvents or extended storage conditions To ensure optimal performance these centrifuge bottles must be filled to at least 80% of their total capacity for proper balance and reliable centri