Learn More
Alkali Scientific CellPro™ Premium Cell Strainers, 100µm Yellow, with 1 reducing adapter, Sterile, Individually Wrapped, 50/cs
SDP
Catalog No. NC1810211
Encompass_Preferred
CellPro™ Premium Cell Strainer are a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in 100um (yellow) porosity. The advanced frame/housing design contributes to superior performance and reduced processing time by eliminating the “air-lock” effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers. 50/pk
Catalog No. NC1810211 Supplier Alkali Scientific Supplier No. MT4100
CellPro™ Premium Cell Strainer are a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in 100um (yellow) porosity. The advanced frame/housing design contributes to superior performance and reduced processing time by eliminating the “air-lock” effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers. 50/pk