Learn More
Alkali Scientific CellPro™ Premium Cell Strainers, 70µm, White, with 1 Reducing Adapter, Sterile, Individually Wrapped, 50/cs
SDP
Catalog No. 502336035
Encompass_Preferred
CellPro™ Premium Cell Strainer are a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in 70um (white) porosity. The advanced frame/housing design contributes to superior performance and reduced processing time by eliminating the “air-lock” effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers. 50/pk
Catalog No. 50-233-6035 Supplier Alkali Scientific Supplier No. MT4070
CellPro™ Premium Cell Strainer are a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in 70um (white) porosity. The advanced frame/housing design contributes to superior performance and reduced processing time by eliminating the “air-lock” effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers. 50/pk