Learn More
Alkali Scientific CellPro Reagent Reservoir with
SDP
Supplier:  Alkali Scientific RRN60
Encompass_Preferred
The 60mL Non-Sterile Reagent Reservoir are a valuable addition to any laboratory setting Each pack contains 5 reservoirs with a case comprising 25 packs The inclusion of caps ensures secure storage of reagents and prevents contamination These reservoirs are designed to provide convenience during pipetting procedures allowing for easy access to reagents while minimizing wastage Their non-sterile nature makes them ideal for various applications that do not require sterile conditions saving time and resources in non-sensitive experiments The transparent material of the reservoirs facilitates clear visibility of the reagent volume aiding in precise measurements and reducing errors Whether youre working on sample preparations dilutions or other laboratory tasks these reagent reservoirs offer efficiency and reliability V-shape vottom use with single or multi-channel pipettes Reusable autoclavable Specially designed lid to prevent contamination Clear graduations DNase/RNase free USP V
Catalog No. 50-302-9293