Learn More
Alkali Scientific CellPro™ Red Biohazard Bags 40 X 46", 100/Cs
SDP
Catalog No. 503028755
Encompass_Preferred
Alkali Scientific's Biohazard Bags are made of Linear Low Density material, designed to safely contain hazardous items in hospitals, laboratories, and healthcare facilities. Biohazard symbol with English and Spanish warnings. Puncture resistant. Flat seal bottom. Flat packed in carton. Compatible Trash Cans:44 Gallon35 Gallon Hands-Free
Catalog No. 50-302-8755 Supplier Alkali Scientific Supplier No. KE1145
Alkali Scientific's Biohazard Bags are made of Linear Low Density material, designed to safely contain hazardous items in hospitals, laboratories, and healthcare facilities. Biohazard symbol with English and Spanish warnings. Puncture resistant. Flat seal bottom. Flat packed in carton. Compatible Trash Cans:44 Gallon35 Gallon Hands-Free