Learn More
Alkali Scientific CellPro™ Solution Bottles, 2000mL (2L), Individually Wrapped, Sterile, 24/cs
SDP
Catalog No. 502335972
Encompass_Preferred
These Solution Bottles offer a wide and easy access bottle opening for quick access and non-drip pouring/decanting. The are graduated in 25mL molded increments for accurate filling, and have light weight yet durable wall construction. They come single packed sterile bottle and single packed sterile cap.
Catalog No. 50-233-5972 Supplier Alkali Scientific Supplier No. VR2000
These Solution Bottles offer a wide and easy access bottle opening for quick access and non-drip pouring/decanting. The are graduated in 25mL molded increments for accurate filling, and have light weight yet durable wall construction. They come single packed sterile bottle and single packed sterile cap.