Learn More
Alkali Scientific CellPro™ Reservoir Bottle, Round, 500mL, Sterile, 24/cs
SDP
Catalog No. 502336116
Encompass_Preferred
CellPro™ Reservoir Bottles are light weight yet have durable wall construction. They offer a wide and easy access bottle opening for quick access and non-drip pouring/decanting. They are non-pyrogenic, polystyrene (PS) bottles that are packaged sterile, and individually wrapped. CellPro™ Reservoir Bottles are graduated in 25mL molded increments for accurate filling.
Catalog No. 50-233-6116 Supplier Alkali Scientific Supplier No. VR500
CellPro™ Reservoir Bottles are light weight yet have durable wall construction. They offer a wide and easy access bottle opening for quick access and non-drip pouring/decanting. They are non-pyrogenic, polystyrene (PS) bottles that are packaged sterile, and individually wrapped. CellPro™ Reservoir Bottles are graduated in 25mL molded increments for accurate filling.