Learn More
Alkali Scientific CellPro™ Reservoir Microplates without Caps, Multi Well, 22mL, 12 Channel, Medium Profile 44.5mm Height, 50/Cs, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503029094
Encompass_Preferred
The 22mL reservoir microplates are made of polypropylene (PP), a transparent polymer material with good chemical compatibility for the storage of most polar organic solutions, acidic and alkaline solutions, and other laboratory liquids. • SBS International Standard• Medical grade polypropylene (PP).• Autoclavable at 121 °C/ 15psi.• Non – pyrogen, DNase/RNase Free• Lot traceability• Low and Medium profile designs to satisfy your liquid handling needs• They come without caps, 1/pk, 5/cs
Catalog No. 50-302-9094 Supplier Alkali Scientific Supplier No. RMPN1222
The 22mL reservoir microplates are made of polypropylene (PP), a transparent polymer material with good chemical compatibility for the storage of most polar organic solutions, acidic and alkaline solutions, and other laboratory liquids. • SBS International Standard• Medical grade polypropylene (PP).• Autoclavable at 121 °C/ 15psi.• Non – pyrogen, DNase/RNase Free• Lot traceability• Low and Medium profile designs to satisfy your liquid handling needs• They come without caps, 1/pk, 5/cs