Learn More
Alkali Scientific CellPro™ Reservoir Microplates without Caps, Single Well, 290mL, 12 Channel, Medium Profile 44.5mm Height, 50/Cs, Sterile
SDP
Catalog No. 503029288
Encompass_Preferred
The 290mL reservoir microplates are made of polypropylene (PP), a transparent polymer material with good chemical compatibility for the storage of most polar organic solutions, acidic and alkaline solutions, and other laboratory liquids. Our reservoirs have the standard SBS size, which Is suitable for all kinds of automatic liquid handling systems. The built-in groove at the bottom of the reservoirs can reduce the dead volume of reagent and ensure the consistency of the liquid volume being handled. • SBS International Standard• Medical grade polypropylene (PP).• Autoclavable at 121 °C/ 15psi.• Non – pyrogen, DNase/RNase Free• Lot traceability• Low and Medium profile designs to satisfy your liquid handling needs
Catalog No. 50-302-9288 Supplier Alkali Scientific Supplier No. RMPN12290S
The 290mL reservoir microplates are made of polypropylene (PP), a transparent polymer material with good chemical compatibility for the storage of most polar organic solutions, acidic and alkaline solutions, and other laboratory liquids. Our reservoirs have the standard SBS size, which Is suitable for all kinds of automatic liquid handling systems. The built-in groove at the bottom of the reservoirs can reduce the dead volume of reagent and ensure the consistency of the liquid volume being handled. • SBS International Standard• Medical grade polypropylene (PP).• Autoclavable at 121 °C/ 15psi.• Non – pyrogen, DNase/RNase Free• Lot traceability• Low and Medium profile designs to satisfy your liquid handling needs