Learn More
Alkali Scientific CellPro Reservoir Microplates
SDP
Catalog No. 503028270
Encompass_Preferred
The 15mL reservoir microplates are designed without caps featuring a multi-well configuration with 12 channels With a low profile standing at 31 4mm height these plates have a capacity of 15ml They come in a pack of 10 and are conveniently packed in a case of 50 Ideal for organizing samples and reagents during experimental procedures these microplates offer a reliable platform for conducting assays and screenings Additionally the well-thought-out design ensures ease of use and compatibility with various automated liquid handling systems making them a practical choice for research facilities diagnostic laboratories and academic institutions Matched reservoir breaker (for 96 and 384) and the cover can avoid the liquid wastage in the process of operation Can be stored at -80 Low heavy metal content Used high quality imported PP material for high stability ensuring no chemical reactions with test reagents Compatible with DMSO and inert to water Certified DNase/RNase and Pyroge
Catalog No. 50-302-8270 Supplier Alkali Scientific Supplier No. RMPN1215
Add to Cart
Add to Cart
The 15mL reservoir microplates are designed without caps featuring a multi-well configuration with 12 channels With a low profile standing at 31 4mm height these plates have a capacity of 15ml They come in a pack of 10 and are conveniently packed in a case of 50 Ideal for organizing samples and reagents during experimental procedures these microplates offer a reliable platform for conducting assays and screenings Additionally the well-thought-out design ensures ease of use and compatibility with various automated liquid handling systems making them a practical choice for research facilities diagnostic laboratories and academic institutions Matched reservoir breaker (for 96 and 384) and the cover can avoid the liquid wastage in the process of operation Can be stored at -80 Low heavy metal content Used high quality imported PP material for high stability ensuring no chemical reactions with test reagents Compatible with DMSO and inert to water Certified DNase/RNase and Pyroge