Learn More
Alkali Scientific CellPro™ Reservoir Microplates without Caps, Single Well, 195mL, 96 Trough, Low Profile 31.4mm Height, 50/Cs, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503028272
Encompass_Preferred
Our Reservoir Microplates offer a convenient solution for various laboratory applications. These single-well plates feature 96 troughs, providing ample space for samples. With a low profile height of 31.4mm, they are compact yet offer a generous volume of 195ml. The non-sterile feature allows flexibility in handling and usage preferences. Our reservoirs have the standard SBS size, which Is suitable for all kinds of automatic liquid handling systems. The built-in groove at the bottom of the reservoirs can reduce the dead volume of reagent and ensure the consistency of the liquid volume being handled.  
Catalog No. 50-302-8272 Supplier Alkali Scientific Supplier No. RMPN96195
Our Reservoir Microplates offer a convenient solution for various laboratory applications. These single-well plates feature 96 troughs, providing ample space for samples. With a low profile height of 31.4mm, they are compact yet offer a generous volume of 195ml. The non-sterile feature allows flexibility in handling and usage preferences. Our reservoirs have the standard SBS size, which Is suitable for all kinds of automatic liquid handling systems. The built-in groove at the bottom of the reservoirs can reduce the dead volume of reagent and ensure the consistency of the liquid volume being handled.  