Learn More
Alkali Scientific CellPro™ Reservoir Microplates without Caps, Single Well, 290mL, 8 Channel, Medium Profile 44.5mm Height, 50/Cs, Sterile
SDP
Catalog No. 503028335
Encompass_Preferred
The 22mL reservoir microplates are made of polypropylene (PP), a transparent polymer material with good chemical compatibility for the storage of most polar organic solutions, acidic and alkaline solutions, and other laboratory liquids. Our reservoirs have the standard SBS size, which Is suitable for all kinds of automatic liquid handling systems. The built-in groove at the bottom of the reservoirs can reduce the dead volume of reagent and ensure the consistency of the liquid volume being handled. • Matched reservoir breaker (for 96 and 384) and the cover can avoid the liquid wastage in the process of operation.• Can be stored at -80℃• Low heavy metal content• Used high quality, imported PP material for high stability, ensuring no chemical reactions with test reagents.• Compatible with DMSO and inert to water• Certified DNase/RNase and Pyrogen Free• Conformed to ANSI/SBS Standards
Catalog No. 50-302-8335 Supplier Alkali Scientific Supplier No. RMPN8290S
The 22mL reservoir microplates are made of polypropylene (PP), a transparent polymer material with good chemical compatibility for the storage of most polar organic solutions, acidic and alkaline solutions, and other laboratory liquids. Our reservoirs have the standard SBS size, which Is suitable for all kinds of automatic liquid handling systems. The built-in groove at the bottom of the reservoirs can reduce the dead volume of reagent and ensure the consistency of the liquid volume being handled. • Matched reservoir breaker (for 96 and 384) and the cover can avoid the liquid wastage in the process of operation.• Can be stored at -80℃• Low heavy metal content• Used high quality, imported PP material for high stability, ensuring no chemical reactions with test reagents.• Compatible with DMSO and inert to water• Certified DNase/RNase and Pyrogen Free• Conformed to ANSI/SBS Standards