Learn More
Alkali Scientific CellPro™ RIPA Buffer + Triton + Glycerol, 2X, 250mL
SDP
Catalog No. 502389265
Encompass_Preferred
Alkali Scientific™ CellPro™ RIPA Buffer + Triton + Glycerol is offered in a convenient 2X concentration, ensuring optimal performance for your protein extraction and solubilization needs. With a sterile and filtered formulation, this buffer guarantees the purity and integrity required for sensitive biochemical assays and downstream applications. RIPA buffer, combined with Triton X-100 and glycerol, facilitates efficient disruption of cells and solubilization of proteins, making it ideal for a wide range of laboratory protocols, including Western blotting, immunoprecipitation, and protein-protein interaction studies. The precisely calibrated composition of CellPro™ RIPA Buffer ensures consistent and reliable results, empowering researchers to achieve accurate and reproducible data in their experiments. Trust in the quality and reliability of CellPro™ RIPA Buffer + Triton + Glycerol to enhance the efficiency and effectiveness of your protein analysis workflows.
Catalog No. 50-238-9265 Supplier Alkali Scientific Supplier No. DB0240
Alkali Scientific™ CellPro™ RIPA Buffer + Triton + Glycerol is offered in a convenient 2X concentration, ensuring optimal performance for your protein extraction and solubilization needs. With a sterile and filtered formulation, this buffer guarantees the purity and integrity required for sensitive biochemical assays and downstream applications. RIPA buffer, combined with Triton X-100 and glycerol, facilitates efficient disruption of cells and solubilization of proteins, making it ideal for a wide range of laboratory protocols, including Western blotting, immunoprecipitation, and protein-protein interaction studies. The precisely calibrated composition of CellPro™ RIPA Buffer ensures consistent and reliable results, empowering researchers to achieve accurate and reproducible data in their experiments. Trust in the quality and reliability of CellPro™ RIPA Buffer + Triton + Glycerol to enhance the efficiency and effectiveness of your protein analysis workflows.