Learn More
Alkali Scientific CellPro™ RPMI 1640 medium with HEPES, L-glutamine, 500ml, 10/Case
SDP
Catalog No. 503027088
Encompass_Preferred
Supports cell growth in research and biomanufacturing cell culture processes. RPMI 1640 medium is extensively quality control tested to ensure lot-to-lot consistency. Medium efficacy and homogeneity of RPMI 1640 is tested. RPMI solution with L-glutamine is available with or without HEPES. RPMI 1640 formulations without L-glutamine as well as RPMI lyophilized powder are also available. RPMI 1640 culture medium is hydrated using purified process water and is filtered through a 0.1 µm filte
Catalog No. 50-302-7088 Supplier Alkali Scientific Supplier No. DR3605CS
Add to Cart
Add to Cart
Supports cell growth in research and biomanufacturing cell culture processes. RPMI 1640 medium is extensively quality control tested to ensure lot-to-lot consistency. Medium efficacy and homogeneity of RPMI 1640 is tested. RPMI solution with L-glutamine is available with or without HEPES. RPMI 1640 formulations without L-glutamine as well as RPMI lyophilized powder are also available. RPMI 1640 culture medium is hydrated using purified process water and is filtered through a 0.1 µm filte