Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Flat Bottom, PTFE (Teflon) Faced Liner, 15mL, 16x125mm, 250/Pk
SDP
Catalog No. 5000662699
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are designed to support dependable sample handling, storage, and culturing in microbiology, clinical diagnostics, and laboratory research environments. Constructed from high quality glass, these tubes feature durable phenolic screw caps that provide a secure, leak resistant seal while tolerating repeated sterilization cycles and routine laboratory handling. The flat bottom design allows the tubes to stand upright on laboratory benches and work surfaces without additional racks, improving stability during filling and labeling processes. An integrated white marking area allows clear sample identification and traceability, helping maintain organized workflows. Compatible with standard laboratory racks and accessories, these tubes are ideal for culturing, reagent storage, and sample preparation. Each tube includes a PTFE (Teflon) faced liner to enhance chemical resistance and sealing performance.
Catalog No. 50-006-62699 Supplier Alkali Scientific Supplier No. STSC15PTFEF
CellPro™ Screw Cap Culture Tubes are designed to support dependable sample handling, storage, and culturing in microbiology, clinical diagnostics, and laboratory research environments. Constructed from high quality glass, these tubes feature durable phenolic screw caps that provide a secure, leak resistant seal while tolerating repeated sterilization cycles and routine laboratory handling. The flat bottom design allows the tubes to stand upright on laboratory benches and work surfaces without additional racks, improving stability during filling and labeling processes. An integrated white marking area allows clear sample identification and traceability, helping maintain organized workflows. Compatible with standard laboratory racks and accessories, these tubes are ideal for culturing, reagent storage, and sample preparation. Each tube includes a PTFE (Teflon) faced liner to enhance chemical resistance and sealing performance.