Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Flat Bottom, PTFE (Teflon) Faced Liner, 5mL, 12X75mm, 250/Pk
SDP
Catalog No. 5000662796
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are designed for reliable sample handling, storage, and culturing in microbiology, clinical, and laboratory research environments. Manufactured from high quality glass, these tubes include durable phenolic screw caps that provide a secure, leak resistant seal and withstand repeated sterilization cycles and routine laboratory handling. The flat bottom design allows tubes to stand upright on benches and work surfaces without requiring additional racks, improving stability during filling, labeling, and storage. An integrated white marking area enables clear sample identification and traceability for organized workflows. These tubes are compatible with standard laboratory racks and accessories and are suitable for culturing, reagent storage, and sample preparation. Each tube includes a PTFE (Teflon) faced liner that provides excellent chemical resistance and enhanced sealing performance.
Catalog No. 50-006-62796 Supplier Alkali Scientific Supplier No. STSC05PTFEF
CellPro™ Screw Cap Culture Tubes are designed for reliable sample handling, storage, and culturing in microbiology, clinical, and laboratory research environments. Manufactured from high quality glass, these tubes include durable phenolic screw caps that provide a secure, leak resistant seal and withstand repeated sterilization cycles and routine laboratory handling. The flat bottom design allows tubes to stand upright on benches and work surfaces without requiring additional racks, improving stability during filling, labeling, and storage. An integrated white marking area enables clear sample identification and traceability for organized workflows. These tubes are compatible with standard laboratory racks and accessories and are suitable for culturing, reagent storage, and sample preparation. Each tube includes a PTFE (Teflon) faced liner that provides excellent chemical resistance and enhanced sealing performance.