Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Flat Bottom, PTFE (Teflon) Faced Liner, 9mL, 13x100mm, 250/Pk
SDP
Catalog No. 5000663123
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes are engineered for dependable performance in microbiology, clinical diagnostics, and laboratory research applications. Constructed from high quality laboratory glass, these tubes feature durable phenolic screw caps that create a tight, leak resistant seal and tolerate repeated sterilization and routine laboratory use. The flat bottom design allows the tubes to stand upright on laboratory benches and work surfaces without the need for additional racks, improving stability during filling and labeling procedures. A clearly defined white marking area enables accurate sample identification and traceability, supporting organized workflows in busy laboratories. Compatible with standard laboratory racks and accessories, these tubes are suitable for culturing, reagent storage, and sample preparation. Each tube includes a PTFE (Teflon) faced liner that improves sealing integrity and chemical resistance.
Catalog No. 50-006-63123 Supplier Alkali Scientific Supplier No. STSC09PTFEF
CellPro™ Screw Cap Culture Tubes are engineered for dependable performance in microbiology, clinical diagnostics, and laboratory research applications. Constructed from high quality laboratory glass, these tubes feature durable phenolic screw caps that create a tight, leak resistant seal and tolerate repeated sterilization and routine laboratory use. The flat bottom design allows the tubes to stand upright on laboratory benches and work surfaces without the need for additional racks, improving stability during filling and labeling procedures. A clearly defined white marking area enables accurate sample identification and traceability, supporting organized workflows in busy laboratories. Compatible with standard laboratory racks and accessories, these tubes are suitable for culturing, reagent storage, and sample preparation. Each tube includes a PTFE (Teflon) faced liner that improves sealing integrity and chemical resistance.